ContactUsLinkComponent
Capping tool, Cap-It
Capping tool, Cap-It
  732-4817
undefined
Capping tool, Cap-It
  732-4817
  • Description:
    Capping tool, Cap-It
  • Pk:
    1
Designed to assist users in applying domed caps to tube strips and plates.
  • Minimises the pressure required to seal the caps effectively
  • Enables multiple tubes to be sealed simultaneously