ContactUsLinkComponent
Sealing tape, light blocking, non-fluorescent, Nunc™
Sealing tape, light blocking, non-fluorescent, Nunc™
Catalog # 731-0750
   Low Manufacturing Impact
undefined
Sealing tape, light blocking, non-fluorescent, Nunc™
Catalog # 731-0750
   Low Manufacturing Impact
Specifications
  • Optical property:
    Light blocking
  • Description:
    Sealing tape, light-blocking
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    50
Specifications
About this item
Vinyl film with pressure sensitive acrylate adhesive. For application to plate top or bottom, to protect light sensitive samples.
  • Temperature stable from −40 to +80 °C
  • Non-pierceable by pipette
Colour: black
External L×W: 123×79 mm