ContactUsLinkComponent
Roller for adhesive seal application
Roller for adhesive seal application
Catalog # 293-1411
Supplier:  Simport Scientific
undefined
Roller for adhesive seal application
Catalog # 293-1411
Supplier:  Simport Scientific
Specifications
  • Description:
    Roller, rubber, Amplate™
  • Pk:
    1
Specifications
About this item
Ensure a perfect seal when using either SecureSeal™ Thermal sealing film or foil on PCR plates. Roller made of medium-hard rubber.
  • Heavy-duty handle with comfortable grip reducing fatigue
  • Durable construction