Orders LinkContactUsLinkComponent
Adhesive seals
Adhesive seals
   Sustainable Options Available
Supplier:   Greiner Bio-One
Adhesive seals
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
Adhesive seals
2 Options Available Below
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
  • Free from RNase, DNase and human DNA
  • Non pyrogenic
Specifications for Product Family
Specifications
  • Catalog No:
  • GREI676002_200
  • 736-0127
  • GREI676001BE
  • GREI676050UK
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
  • Optical property
    Sterile
    Availability
    Price
  • GREI676002_200
    EASYseal adhesive film, non sterile, 120×80 mm
    EASYseal adhesive film, non sterile, 120×80 mm
    Transparent
    -
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 73,50 €
    73,50 €
    1 * 200 ST
       73,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    GREI676002_200
    Environmentally Preferable
    Environmental certification
  • 736-0127
    EASYseal adhesive film, non sterile, 80×140 mm
    EASYseal adhesive film, non sterile, 80×140 mm
    Transparent
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,40 €
    51,40 €
    1 * 100 ST
       51,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    736-0127
    Environmentally Preferable
    Environmental certification
  • GREI676001BE
    EASYseal adhesive film, non sterile, 80×140 mm
    EASYseal adhesive film, non sterile, 80×140 mm
    Transparent
    -
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    GREI676001BE
    Environmentally Preferable
    Environmental certification
  • GREI676050UK
    BREATHseal adhesive film, non sterile
    BREATHseal adhesive film, non sterile
    Transparent
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI676050UK
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact