Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   BOCHEM
Scoops
Supplier:  BOCHEM
Certificates
Scoops
5 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling, anti-magnetic
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 120 ml
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,50 €
    19,50 €
    1 * 1 ST
       19,50 €
  • 450 ml
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,90 €
    21,90 €
    1 * 1 ST
       21,90 €
  • 1000 ml
    320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,70 €
    25,70 €
    1 * 1 ST
       25,70 €
  • 1500 ml
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,80 €
    29,80 €
    1 * 1 ST
       29,80 €
  • 2000 ml
    370 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,70 €
    41,70 €
    1 * 1 ST
       41,70 €