Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   NICKEL ELECTRO
Scoops
Supplier:  NICKEL ELECTRO
Certificates
Scoops
4 Options Available Below
Product Details & Documents
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
  • Excellent thermal resistance, autoclavable
  • Non magnetic, bright finish
  • Highly polished surface makes cleaning easier
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 500 ml
    325 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,00 €
    56,00 €
    1 * 1 ST
       56,00 €
  • 100 ml
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,30 €
    36,30 €
    1 * 1 ST
       36,30 €
  • 250 ml
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,20 €
    43,20 €
    1 * 1 ST
       43,20 €
  • 1000 ml
    340 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,70 €
    60,70 €
    1 * 1 ST
       60,70 €