Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   NICKEL ELECTRO
Scoops
Supplier:  NICKEL ELECTRO
Certificates
Scoops
4 Options Available Below
Product Details & Documents
Stainless steel. Round bottom. Ideal for use as weighing boats etc.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Product Family Options
Product Information
  • Capacity
    Length
    Width
    Availability
    Price
  • 25 ml
    70 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 63,60 €
    63,60 €
    1 * 5 ST
       63,60 €
  • 32 ml
    90 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 65,30 €
    65,30 €
    1 * 5 ST
       65,30 €
  • 37 ml
    105 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 65,30 €
    65,30 €
    1 * 5 ST
       65,30 €
  • 42 ml
    120 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 67,20 €
    67,20 €
    1 * 5 ST
       67,20 €