Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   USBECK Laborgeräte
Scoops
Certificates
Scoops
8 Options Available Below
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 60 ml
    155 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,20 €
    4,20 €
    1 * 1 ST
       4,20 €
  • 130 ml
    210 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,90 €
    6,90 €
    1 * 1 ST
       6,90 €
  • 250 ml
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,40 €
    8,40 €
    1 * 1 ST
       8,40 €
  • 450 ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,60 €
    13,60 €
    1 * 1 ST
       13,60 €
  • 800 ml
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,20 €
    23,20 €
    1 * 1 ST
       23,20 €
  • 1500 ml
    390 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,00 €
    35,00 €
    1 * 1 ST
       35,00 €
  • 2500 ml
    410 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,30 €
    45,30 €
    1 * 1 ST
       45,30 €
  • 375 ml
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €