Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   RSG
Scoops
Supplier:  RSG
Certificates
Scoops
5 Options Available Below
Product Details & Documents
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design
Documents
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 120 ml
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,00 €
    22,00 €
    1 * 1 ST
       22,00 €
  • 450 ml
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,00 €
    25,00 €
    1 * 1 ST
       25,00 €
  • 1000 ml
    320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €
  • 1500 ml
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • 2500 ml
    365 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,50 €
    49,50 €
    1 * 1 ST
       49,50 €