Orders LinkContactUsLinkComponent
Measuring scoops, Azlon®
Measuring scoops, Azlon®
Supplier:   SciLabware
Measuring scoops, Azlon®
Supplier:  SciLabware
Certificates
Measuring scoops, Azlon®
Measuring scoops, Azlon®
7 Options Available Below
Product Details & Documents
PP, white. Scoops with flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, robust
  • Heat resistant up to +130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 250 ml
    260 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 52,60 €
    52,60 €
    1 * 5 ST
       52,60 €
  • 50 ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 16,70 €
    16,70 €
    1 * 5 ST
       16,70 €
  • 10 ml
    100 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 8,10 €
    8,10 €
    1 * 5 ST
       8,10 €
  • 25 ml
    135 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 11,40 €
    11,40 €
    1 * 5 ST
       11,40 €
  • 500 ml
    315 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 82,30 €
    82,30 €
    1 * 5 ST
       82,30 €
  • 1000 ml
    385 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,30 €
    23,30 €
    1 * 1 ST
       23,30 €
  • 100 ml
    198 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 24,60 €
    24,60 €
    1 * 5 ST
       24,60 €