Orders LinkContactUsLinkComponent
Stand bases
Supplier:   USBECK Laborgeräte
Stand bases
Certificates
Stand bases
7 Options Available Below
Product Details & Documents
Stainless steel. Firm, non slip stand for a wide variety of uses. With an M10 threaded hole on face side and adjustable feet for uneven surfaces.
  • Rubber feet for a firm footing
  • Smooth surfaces for easy, fast cleaning
  • Robust
DIN 12892/55
 : DIN 12892/55
Product Family Options
Product Information
  • L×W
    Weight
    Availability
    Price
  • 210×130 mm
    1,7 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,20 €
    85,20 €
    1 * 1 ST
       85,20 €
  • 210×130 mm
    1,7 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,90 €
    36,90 €
    1 * 1 ST
       36,90 €
  • 250×160 mm
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • 300×150 mm
    2,8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,80 €
    52,80 €
    1 * 1 ST
       52,80 €
  • 315×200 mm
    4,0 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • 315×200 mm
    4,0 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,90 €
    57,90 €
    1 * 1 ST
       57,90 €
Plates with 100 cm threading rod
Product Information
  • L×W
    Weight
    Availability
    Price
  • 300×150 mm
    3,7 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 147,00 €
    147,00 €
    1 * 1 ST
       147,00 €