Orders LinkContactUsLinkComponent
VWR®, Electrode stand
VWR®, Electrode stand
Electrode stand, h-shaped, 2 electrodes
Catalog # 241-0114
Supplier:   VWR Collection
VWR®, Electrode stand
Catalog # 241-0114
Supplier:  VWR Collection
VWR®, Electrode stand
VWR®, Electrode stand
Electrode stand, h-shaped, 2 electrodes
Price
159,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Grey iron stand for two electrodes.
  • Sturdy painted cast iron stand, provides a firm footing
  • Stainless steel support rod
  • Acrylic electrode holder fitted using knurling screws
Specifications
  • For:
    2 electrodes
  • Description:
    Electrode stand, h-shaped