Orders LinkContactUsLinkComponent
Stand bases
Stand bases
Supplier:   R L ENTERPRISES
Stand bases
Supplier:  R L ENTERPRISES
Certificates
Stand bases
8 Options Available Below
Product Details & Documents
Rectangular retort bases with M10 threaded hole in centre of short side.
  • Steel, with acrylic finish
  • Acid resistant surface
  • Non-slip rubber feet for a firm footing
Ordering information: Supplied without rods which need to be ordered separately.
Product Family Options
Product Information
  • L×W
    Weight
    Colour
    Availability
    Price
  • 100×160 mm
    0,75 kg
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,40 €
    51,40 €
    1 * 1 ST
       51,40 €
  • 100×160 mm
    0,75 kg
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,30 €
    50,30 €
    1 * 1 ST
       50,30 €
  • 125×200 mm
    1,25 kg
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,60 €
    55,60 €
    1 * 1 ST
       55,60 €
  • 125×200 mm
    1,25 kg
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,40 €
    53,40 €
    1 * 1 ST
       53,40 €
  • 160×250 mm
    2 kg
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,10 €
    80,10 €
    1 * 1 ST
       80,10 €
  • 160×250 mm
    2 kg
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 78,30 €
    78,30 €
    1 * 1 ST
       78,30 €
  • 200×315 mm
    3 kg
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 200×315 mm
    3 kg
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,10 €
    92,10 €
    1 * 1 ST
       92,10 €
Retort rods, stainless steel
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 750 mm
    12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,70 €
    81,70 €
    1 * 1 ST
       81,70 €
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 500 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,90 €
    43,90 €
    1 * 1 ST
       43,90 €
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 1000 mm
    12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
Retort rods, zinc plated steel
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 500 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 222,00 €
    222,00 €
    1 * 10 ST
       222,00 €
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 600 mm
    12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,30 €
    27,30 €
    1 * 1 ST
       27,30 €
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 750 mm
    12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 1000 mm
    12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,80 €
    35,80 €
    1 * 1 ST
       35,80 €