Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Labomeca® Rods without Thread

VWR® Labomeca® Rods without Thread

8 mm, 800 mm
Catalog # 241-0066
Supplier:   VWR Collection
VWR® Labomeca® Rods without Thread
Catalog # 241-0066
Supplier:  VWR Collection
VWR® Labomeca® Rods without Thread
VWR® Labomeca® Rods without Thread

8 mm, 800 mm

Price
53,10 €
Log in for availability
1 * 5 ST
Product Details & Documents

Duralumin, solid.

  • Good chemical resistance
  • Solid, lightweight, very durable
Specifications
  • Ø:
    8 mm
  • Length:
    800 mm