Orders LinkContactUsLinkComponent
Funnel holders
Supplier:   KARTELL
Funnel holders
Supplier:  KARTELL
Certificates
Funnel holders
2 Options Available Below
Product Details & Documents
PP, white, unbreakable.
  • For funnels 50 to 120 mm Ø
  • With bosshead to fit rods 8 to 14 mm Ø
  • Interchangeable insert for different sizes
Product Family Options
Product Information
  • For
    Availability
    Price
  • 1 funnel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,50 €
    31,50 €
    1 * 1 ST
       31,50 €
  • 2 funnels
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,30 €
    43,30 €
    1 * 1 ST
       43,30 €