Orders LinkContactUsLinkComponent
O-rings for flat flanges, DURAN®
O-rings for flat flanges, DURAN®
Supplier:   DWK Life Sciences
O-rings for flat flanges, DURAN®
Supplier:  DWK Life Sciences
Certificates
O-rings for flat flanges, DURAN®
O-rings for flat flanges, DURAN®
5 Options Available Below
Product Details & Documents
Silicone (VMQ), transparent.
  • Guarantee excellent joint seal
  • Heat resistant from −50 to +200 °C
  • Good chemical resistance
O-rings for flat flanges with groove.
Product Family Options
Product Information
  • For
    Colour
    Material
    Ø int.
    Availability
    Price
  • Flat flanges DN 60
    Transparent
    Silicone
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,20 €
    13,20 €
    1 * 1 ST
       13,20 €
  • Flat flanges DN 100
    Transparent
    Silicone
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,10 €
    21,10 €
    1 * 1 ST
       21,10 €
  • Flat flanges DN 150
    Transparent
    Silicone
    133 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,80 €
    30,80 €
    1 * 1 ST
       30,80 €
  • Flat flanges DN 150
    Transparent
    Silicone
    157 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,70 €
    41,70 €
    1 * 1 ST
       41,70 €
  • Flat flanges DN 200
    Transparent
    Silicone
    215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,20 €
    53,20 €
    1 * 1 ST
       53,20 €