Flat flange lids with 4 or 5 necks, Quickfit®
Supplier: SciLabware
Certificates
About this item
Lids, PYREX® borosilicate glass, with precision ground, flat flange joints.
- For use with Quickfit® reaction flasks and culture vessels
- Excellent chemical resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Frequently Bought Together