Orders LinkContactUsLinkComponent
Racks, Plexiglas for tubes
 :   KARTELL
Racks, Plexiglas for tubes
 :  KARTELL
Racks, Plexiglas for tubes
Racks, Plexiglas for tubes
Plexiglas (PMMA)
  • Two tiers
  • Smaller holes in the lower tier to hold test tubes securely
Product Family Options
  • No. of Holes
    For tube Ø
    W×D×H
  • 12 (2×6)
    28 mm
    255×90×80 mm
    1 * 1 ST
    1 * 1 ST - 78,70 €
    78,70 €
    1 * 1 ST
       78,70 €
  • 12 (2×6)
    35 mm
    255×90×80 mm
    1 * 1 ST
    1 * 1 ST - 77,70 €
    77,70 €
    1 * 1 ST
       77,70 €
  • 12 (2×6)
    40 mm
    310×110×80 mm
    1 * 1 ST
    1 * 1 ST - 85,00 €
    85,00 €
    1 * 1 ST
       85,00 €