Orders LinkContactUsLinkComponent
Racks for test tubes
Racks for test tubes
Supplier:   HUPFER METALLWERKE
Racks for test tubes
Supplier:  HUPFER METALLWERKE
Certificates
Racks for test tubes
Racks for test tubes
14 Options Available Below
Product Details & Documents
Stainless steel, rust-proof, electrolytically or steel polished, polyamide coated.
  • Excellent chemical resistance
  • Excellent thermal resistance (polyamide coating resistant up to 80 °C)
  • Robust, stable construction and easy to clean
Wire racks with second level tier, 21 mm high. Total height 68 mm. Hole size 18×18 mm.
Product Family Options
Product Information
  • No. of Holes
    Material
    W×D×H
    Availability
    Price
  • 12 (2×6)
    Stainless steel
    130×50×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,60 €
    65,60 €
    1 * 1 ST
       65,60 €
  • 18 (3×6)
    Stainless steel
    130×70×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 24 (2×12)
    Stainless steel
    250×50×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • 24 (4×6)
    Stainless steel
    125×85×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,50 €
    92,50 €
    1 * 1 ST
       92,50 €
  • 36 (3×12)
    Stainless steel
    250×70×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 120,00 €
    120,00 €
    1 * 1 ST
       120,00 €
  • 48 (4×12)
    Stainless steel
    250×90×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • 100 (10×10)
    Steel, PA coated
    210×210×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €
  • 100 (5×20)
    Steel, PA coated
    410×180×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • 48 (4×12)
    Steel, PA coated
    250×90×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 12 (2×6)
    Steel, PA coated
    130×50×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,10 €
    66,10 €
    1 * 1 ST
       66,10 €
  • 18 (3×6)
    Steel, PA coated
    130×70×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,10 €
    75,10 €
    1 * 1 ST
       75,10 €
  • 24 (2×12)
    Steel, PA coated
    250×50×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,70 €
    95,70 €
    1 * 1 ST
       95,70 €
  • 24 (4×6)
    Steel, PA coated
    130×90×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,80 €
    91,80 €
    1 * 1 ST
       91,80 €
  • 36 (3×12)
    Steel, PA coated
    250×70×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,00 €
    98,00 €
    1 * 1 ST
       98,00 €