Orders LinkContactUsLinkComponent
Staining racks
Staining racks
Staining rack
Catalog # 720-0096
Supplier:   DIXON SCIENTIFIC
Staining racks
Catalog # 720-0096
Supplier:  DIXON SCIENTIFIC
Staining racks
Staining racks
Staining rack
Price
422,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Comprises two brass end-plates fitted with levelling screws and clips.
  • For holding two glass rods with diameter of ∼8 to 9 mm
Ordering information: Rods are not included.
Specifications
  • Description:
    Staining rack