Order Entry
Belgium
Orders LinkContactUsLinkComponent
Staining racks

Staining racks

Staining rack, 86×695×21 mm
Catalog # 631-9520
Supplier:   KARTELL
Staining racks
Catalog # 631-9520
Supplier:  KARTELL
Staining racks
Staining racks

Staining rack, 86×695×21 mm

Price
16,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP.

  • For use with 631-9511

Staining rack with horizontal tray, holds 20 standard slides 76×26 mm.

W×D×H: 86×695×21 mm

Specifications
  • Description:
    Staining rack
  • W×D×H:
    86×695×21 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.