ContactUsLinkComponent
Racks for microcentrifuge tubes
Racks for microcentrifuge tubes
Catalog # HEATHS4284A
Supplier:  HEATHROW
undefined
Racks for microcentrifuge tubes
Catalog # HEATHS4284A
Supplier:  HEATHROW
Specifications
  • For tubes:
    1,5/2,0 ml
  • No. of Holes:
    20
  • Colour:
    Blue
  • W×D×H:
    212×89×51 mm
  • Pk:
    6
Specifications
About this item
PP, autoclavable.
  • Racks lock together on each side permitting the user to attach several for larger experiments
  • “Hold-it” finger slot on the front
  • Internal ribbing for stability