Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Draining rack, wall mounted

Draining rack, 450×110×630 mm
  149-1310
 :   KARTELL
Draining rack, wall mounted
  149-1310
 :  KARTELL
Draining rack, wall mounted
Draining rack, wall mounted

Draining rack, 450×110×630 mm

 
216,00 €
   
1 * 1 ST
 

PS, gray.

  • The draining rods can be inserted and replaced as required
  • Supplied complete with drainage tube and fixtures for wall mounting
  • Optional rods for narrow-necked vessels are available to order under article number 212-6109

With integral extra wide draining channel and 72 draining rods which are 95 mm long, 15 mm Ø. The rod insertion apertures are closed at the back. This prevents water from seeping into the wall.

 
  • Description:
    Draining rack
  • W×D×H:
    450×110×630 mm
Accessories

 

  •  
    Description
    Ø×L
     
     
  • 212-6109
    Small pegs for draning rack
    6×95 mm
       
    1 * 11 ST - 28,10 €

 

  •  
    Description
    Ø×L
     
     
  • KART21308
    Standard pegs for draning rack
    15×95 mm
       
    1 * 72 ST - 97,20 €