Orders LinkContactUsLinkComponent
Draining racks, Azlon®
Draining racks, Azlon®
Supplier:   SciLabware
Draining racks, Azlon®
Supplier:  SciLabware
Certificates
Draining racks, Azlon®
Draining racks, Azlon®
2 Options Available Below
Product Details & Documents
Easy to assemble, lightweight polystyrene draining racks designed to be wall mounted.
  • Pegs slot into place on the back board and are spaced to allow various sizes of lab ware to fit on to the rack
  • Water is collected in the trough at the base of the rack and drains away via a rubber tube which can be directed into a nearby sink or container
Ø/peg: 16 mm
Delivery information: Supplied with wall mounting accessories.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 212-7548
    Rack with 72 pegs
    Rack with 72 pegs
    450×120×650 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 324,00 €
    324,00 €
    1 * 1 ST
       324,00 €
  • AZLOWRP002P
    Set of 11 narrow pegs
    Set of 11 narrow pegs
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 57,30 €
    57,30 €
    1 * 1 SET
       57,30 €