Order Entry
Belgium
Orders LinkContactUsLinkComponent
Draining rack, Azlon®

Draining rack, Azlon®

Supplier:   SciLabware
Draining rack, Azlon®
Supplier:  SciLabware
Certificates
Draining rack, Azlon®
Draining rack, Azlon®

2 Options Available Below

Product Details & Documents

Wire, epoxy coated.

  • Eight pegs 110 mm and 24 pegs 80 mm long
  • Available either wall mountable or with free-standing support for benchtop use

High quality draining rack made from chemically resistant epoxy coated steel. Suitable for use with plastic and glass labware.

Product Family Options

Product Information

  • W×D×H
    Availability
    Price
  • 212-7543
    Rack only
    Rack only
    300×110×410 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €

  • 212-7544
    Rack with integral free standing support
    Rack with integral free standing support
    300×110×410 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.