Orders LinkContactUsLinkComponent
Vial racks, WHEATON®
Vial racks, WHEATON®
Vial racks, WHEATON®
Vial racks, WHEATON®
These polypropylene racks accommodate 36 or 50 headspace auto-sampler vials. Each well has an opening in the bottom to facilitate drainage.
  • Autoclavable and stackable
  • Chemically resistant
  • Include position identifiers
Product Family Options
  • Hole Ø
    No. of Holes
    W×D×H
  • 23,1 mm
    36
    322×91×28 mm
    1 * 5 ST
    1 * 5 ST - 253,00 €
    253,00 €
    1 * 5 ST
       253,00 €
  • 12,5 mm
    50
    190×100×22 mm
    1 * 5 ST
    1 * 5 ST - 257,00 €
    257,00 €
    1 * 5 ST
       257,00 €