Racks for centrifuge tubes, Poxygrid®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Epoxy coated steel wire, autoclavable.
- Ideal for supporting conical bottom centrifuge tubes
- The bottom rails hold the tip of the conical tube
Hole Ø: 30 mm, for 525-9100: 18 mm
Frequently Bought Together