Density bottles, adjusted
Supplier: Lenz Laborglas
Aller à :
À propos de cet article
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
- The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
- The top of stopper is ground and polished for ease of wiping clean
- Stopper is size NS 10/19 with capillary
* DIN ISO 3507
5 Options disponibles ci-dessous
Product Details & Documents
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
- The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
- The top of stopper is ground and polished for ease of wiping clean
- Stopper is size NS 10/19 with capillary
* DIN ISO 3507
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Product Family Options
Informations sur le produit
- Volume (nom.)AvailabilityPrice