Twin trough chambers
Supplier: CAMAG
Jump to:
About this item
Twin-trough chambers offer several ways to improve the results of TLC/HPTLC developing techniques.
- Low solvent consumption
- Reproducible pre-equilibration of the plate with solvent vapour is possible by placing it in the empty trough opposite that containing the solvent
Development only starts when the solvent is introduced into the trough with the plate.
* ADC: automatic developing chamber.
7 Options Available Below
Product Details & Documents
Twin-trough chambers offer several ways to improve the results of TLC/HPTLC developing techniques.
- Low solvent consumption
- Reproducible pre-equilibration of the plate with solvent vapour is possible by placing it in the empty trough opposite that containing the solvent
Development only starts when the solvent is introduced into the trough with the plate.
* ADC: automatic developing chamber.
Product Family Options
Product Information
- AvailabilityPrice
- 552-0011Chamber for 100×100 mm plates, with stainless steel lid
- 552-0012Chamber for 100×100 mm plates, without lid
- 552-0018Chamber for 200×100 mm plates, with glass lid
- 552-0019Chamber for 200×100 mm plates, with stainless steel lid
- 552-0020Chamber for 200×200 mm plates, with glass lid
- 552-0021Chamber for 200×200 mm plates, with stainless steel lid
- 552-0023Chamber for 200×200 mm plates, without lid