Orders LinkContactUsLinkComponent
Flat bottom chambers
Flat bottom chambers
Supplier:   CAMAG
Flat bottom chambers
Supplier:  CAMAG
Certificates
Flat bottom chambers
5 Options Available Below
Product Details & Documents
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
Product Family Options
Product Information
  • Availability
    Price
  • 552-0010
    Chamber for 100×100 mm plates, without lid
    Chamber for 100×100 mm plates, without lid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
    1 * 1 ST
       167,00 €
  • 552-0009
    Chamber for 100×100 mm plates, with stainless steel lid
    Chamber for 100×100 mm plates, with stainless steel lid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €
  • 552-0015
    Chamber for 200×200 mm plates, with glass lid
    Chamber for 200×200 mm plates, with glass lid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 345,00 €
    345,00 €
    1 * 1 ST
       345,00 €
  • 552-0022
    Chamber for 200×200 mm plates, without lid
    Chamber for 200×200 mm plates, without lid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 329,00 €
    329,00 €
    1 * 1 ST
       329,00 €
  • 552-0024
    Chamber for 200×200 mm plates, with stainless steel lid
    Chamber for 200×200 mm plates, with stainless steel lid
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 379,00 €
    379,00 €
    1 * 1 ST
       379,00 €
Lids
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Stainless steel lid for 100×100 mm twin trough and flat bottom chambers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,20 €
    36,20 €
    1 * 1 ST
       36,20 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Glass lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,80 €
    20,80 €
    1 * 1 ST
       20,80 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Stainless steel lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,70 €
    56,70 €
    1 * 1 ST
       56,70 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Saturation pads
    Saturation pads
    552-0079
    Log in for availability
    1 * 100 ST - 245,00 €