Order Entry
Belgium
Orders LinkContactUsLinkComponent
TLC sprayer

TLC sprayer

TLC sprayer with 2 spray heads, 1×0,8 and 1×1,25 mm
Catalog # 1.08540.0001
Supplier:   Merck
TLC sprayer
Catalog # 1.08540.0001
Supplier:  Merck
TLC sprayer
TLC sprayer

TLC sprayer with 2 spray heads, 1×0,8 and 1×1,25 mm

Price
974,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The Merck TLC sprayer allows derivatisation reagents to be sprayed homogeneously onto developed chromatograms. It is equipped with two different spray heads, (0,8 and 1,25 mm), that are optimised for low and high viscosity solutions respectively. The electro-pneumatically operated sprayer uses compressed air-driven by accumulator power and inductive charging.

Documents

Specifications
  • Description:
    TLC sprayer with 2 spray heads, 1×0,8 and 1×1,25 mm
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • 1.06705.0100
    100 ml
    Log in for availability
    1 * 100 mL - 77,70 €

Product Information

  • Catalog #
    Pack type
    Pk
    Availability
    Price
  • 1.02035.0100
    Glass bottle
    100 ml
    Log in for availability
    1 * 100 mL - 57,70 €

Product Information

  • Catalog #
    Pack type
    Pk
    Availability
    Price
  • 1.00480.0100
    Glass bottle
    100 mL
    Log in for availability
    1 * 100 mL - 80,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Spray heads for TLC sprayer, 5×0,8 and 1×1,25 mm
    Spray heads for TLC sprayer, 5×0,8 and 1×1,25 mm
    1.08541.0001
    Log in for availability
    1 * 6 ST - 197,00 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.