HPTLC plates with spherical particles, LiChrospher®
Supplier: Merck
Certificates
About this item
HPTLC LiChrospher® plates are based on spherical-shaped silica 60 with a particle size of 7 µm, and narrow particle size distribution similar to the phases used in HPLC. LiChrospher® HPTLC plates possess a selectivity that is comparable to respective HPTLC plates, however, plate height and separation numbers are further improved resulting in shorter analysis times.
- 20% reduced running times
- Highly compact spots or zones
- Lower detection limits and better resolution
LiChrospher® HPTLC plates are especially suitable for separations of highly complex, low concentration samples, such as:
Trace analysis of pesticide mixtures
Assay of pharmaceutical compounds