Robotic tips, Axygen®, for Beckman®
Supplier: Corning
About this item
Free of RNase, DNase, DNA and pyrogens.
- Transparent PP
- Sterile tips: Sterilised by gamma radiation
- Filter tips: PE filter prevents any contamination of the pipette
- Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample
21 Options Available Below
Product Details & Documents
Free of RNase, DNase, DNA and pyrogens.
- Transparent PP
- Sterile tips: Sterilised by gamma radiation
- Filter tips: PE filter prevents any contamination of the pipette
- Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 736-0426
- 736-0425
- 736-0269
- 736-0435
- 736-0436
- 736-0432
- 736-0429
- 736-0430
- 736-0433
- AXYGFX-10-R
- AXYGFX-20-R-S
- AXYGFX-20-R
- AXYGFXF-20-R-S
- 736-0254
- AXYGFX-50-R
- AXYGFX-50-R-S
- AXYGFXF-180-R-S
- AXYGFXF-165-WB-R-S
- AXYGFXF-200-R-S
- AXYGFXF-205-WB-R-S
- AXYGFX-250-R
- AXYGFX-250-R-S
- AXYGFX-255-WB-R
- AXYGFX-384-R
- AXYGFX-384-R-S
- AXYGFX-384-XL-R
- AXYGFX-384-XL-RS
- AXYGFXF-384-LRS
- 736-0275
- 613-3815
- 736-0323
- 736-0322
- 736-0324
- 736-0327
- 736-0325
- 736-0326
- 736-0330
- 736-0328
- 736-0329
- 736-0284
- 736-0290
- 736-0282
- 736-0283
- 736-0289
- 736-0293
- 736-0291
- 736-0292
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Biomek FX/NX / Multimek AP96
Product Information
- Tip styleVolumeSterileFilterTip colourFor use withPackedAvailabilityPrice
- FXF-165-WB-R-S (wide bore)+TransparentAP-96 / SPAN-8 / MP200Discontinued-Specifications:
More
Specifications
Cat. No.736-0269Environmentally PreferableEnvironmental certification - FX-10-R10 µl--TransparentAP-96 / SPAN-8 / MP2050 racks, 96 eachLog in for availability1 * 4.800 ST416,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-10-REnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-20-R-S20 µl+-TransparentAP-96 / SPAN-8 / MP2050 racks, 96 eachLog in for availability1 * 4.800 ST428,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-20-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-20-R20 µl--TransparentAP-96 / SPAN-8 / MP2050 racks, 96 eachLog in for availability1 * 4.800 ST377,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-20-REnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FXF-20-R-S20 µl++TransparentAP-96 / SPAN-8 / MP2050 racks, 96 eachLog in for availability1 * 4.800 ST760,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFXF-20-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FXF-50-R-S30 µl++TransparentAP-96 / SPAN-8 / MP2010 racks, 96 eachLog in for availability1 * 960 ST152,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.736-0254Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-50-R50 µl--TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST402,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-50-REnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-50-R-S50 µl+-TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST482,00 €Specifications:
More
Specifications
Cat. No.AXYGFX-50-R-SEnvironmentally PreferableEnvironmental certification - FXF-180-R-S165 µl++TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST760,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFXF-180-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FXF-165-WB-R-S (wide bore)165 µl++TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST760,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFXF-165-WB-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FXF-200-R-S200 µl++TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST714,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFXF-200-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FXF-205-WB-R-S (wide bore)205 µl++TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST974,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFXF-205-WB-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-250-R250 µl--TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST374,00 €Specifications:
More
Specifications
Cat. No.AXYGFX-250-REnvironmentally PreferableEnvironmental certification - FX-250-R-S250 µl+-TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST428,00 €Specifications:
More
Specifications
Cat. No.AXYGFX-250-R-SEnvironmentally PreferableEnvironmental certification - FX-255-WB-R (wide bore)250 µl--TransparentAP-96 / SPAN-8 / MP20050 racks, 96 eachLog in for availability1 * 4.800 ST374,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-255-WB-REnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact
Biomek FX/NX / Multimek AP384
Product Information
- Tip styleVolumeSterileFilterTip colourFor use withPackedAvailabilityPrice
- FX-384-R0,5 - 30 µl--TransparentAP-38450 racks, 384 eachLog in for availability1 * 19.200 ST4 550,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-384-REnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-384-R-S0,5 - 30 µl+-TransparentAP-38450 racks, 384 eachLog in for availability1 * 19.200 ST4 820,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-384-R-SEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-384-XL-R (Extended)0,5 - 30 µl--TransparentAP-38450 racks, 384 eachLog in for availability1 * 19.200 ST4 820,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.AXYGFX-384-XL-REnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-384-XL-R-S (Extended)0,5 - 30 µl+-TransparentAP-38450 racks, 384 eachLog in for availability1 * 19.200 ST5 120,00 €Specifications:
More
Specifications
Cat. No.AXYGFX-384-XL-RSEnvironmentally PreferableEnvironmental certification - FXF-384-L-R-S, Maxymum Recovery™0,5 - 30 µl++TransparentAP-38450 racks, 384 eachLog in for availability1 * 19.200 STSpecifications:
More
Specifications
Cat. No.AXYGFXF-384-LRSEnvironmentally PreferableEnvironmental certification - FXF-384-XL-R-S (Extended)0,5 - 30 µl++TransparentAP-38410 racks, 384 eachLog in for availability1 * 3.840 ST1 390,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.736-0275Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - FX-384-30FP-R (fine point)0,5 - 30 µl--TransparentAP-38410 racks, 384 eachLog in for availability1 * 3.840 ST909,00 €Specifications:
More
Specifications
Cat. No.613-3815Environmentally PreferableEnvironmental certification