Robotic tips for Zymark® (Presto, RapidPlate, SciClone)
Supplier: Thermo Fisher Scientific
Certificates
About this item
High quality tips, especially intended for research in molecular biology laboratories.
- Transparent PP
- Certified free of RNase, DNase, DNA and pyrogens
- Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
- Sterile tips: Sterilised by gamma radiation
Specifications
- Catalog No:
- 613-1885
- 736-0024
- 737-2001
- 613-1865
- 737-2002
- 613-1886
- 737-2000
- 736-0023
- 736-0102
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)