Orders LinkContactUsLinkComponent
Robotic tips for Tecan®
Robotic tips for Tecan®
   Sustainable Options Available
Robotic tips for Tecan®
   Sustainable Options Available
Certificates
Robotic tips for Tecan®
46 Options Available Below
Product Details & Documents
High quality tips, especially intended for research in molecular biology laboratories.
  • Transparent or black PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • Wide aperture tips: Designed for applications with more viscous samples
Ordering information: Available in boxes of 10 racks or 50 racks (Econopak).
Specifications for Product Family
Specifications
  • Catalog No:
  • 736-0045
  • 736-0058
  • 736-0010
  • 736-0009
  • 736-0066
  • 736-0012
  • 736-0011
  • 736-0006
  • 736-0050
  • 736-0008
  • 736-0007
  • 736-0005
  • MLBP902-262XP
  • MLBP902-261XP
  • MLBP903-033
  • 736-0061
  • 736-0043
  • 736-0049
  • 736-0052
  • 736-0054
  • 736-0056
  • 736-0060
  • 736-0062
  • 736-0064
  • 736-0055
  • 736-0053
  • 736-0051
  • 736-0063
  • 736-0059
  • 736-0042
  • 736-0041
  • 736-0048
  • 736-0047
  • 736-0046
  • 736-0044
  • 736-0057
  • 736-0065
  • 732-1527
  • 732-1528
  • 732-1529
  • 732-1530
  • 732-1531
  • 732-1532
  • 732-1533
  • 732-1534
  • 732-1535
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Tecan® Genesis / MiniPrep (transparent)
Product Information
  • Tip style
    Volume
    Sterile
    Filter
    Tip colour
    Packed
    Availability
    Price
  • 901-262
    20 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 174,00 €
    174,00 €
    1 * 960 ST
       174,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0045
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-262
    200 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 167,00 €
    167,00 €
    1 * 960 ST
       167,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0058
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-261
    200 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 183,00 €
    183,00 €
    1 * 960 ST
       183,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0010
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-021
    175 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 224,00 €
    224,00 €
    1 * 960 ST
       224,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0009
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-262
    1000 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 203,00 €
    203,00 €
    1 * 960 ST
       203,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0066
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-261
    1000 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 221,00 €
    221,00 €
    1 * 960 ST
       221,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0012
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-021
    1000 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 224,00 €
    224,00 €
    1 * 960 ST
       224,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0011
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 901-261
    20 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 188,00 €
    188,00 €
    1 * 960 ST
       188,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0006
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-262
    50 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 174,00 €
    174,00 €
    1 * 960 ST
       174,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0050
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-261
    50 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 188,00 €
    188,00 €
    1 * 960 ST
       188,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0008
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-021
    50 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 239,00 €
    239,00 €
    1 * 960 ST
       239,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0007
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 901-021
    10 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 239,00 €
    239,00 €
    1 * 960 ST
       239,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0005
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-262 PATCH XPRESS
    50 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 180,00 €
    180,00 €
    1 * 960 ST
       180,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MLBP902-262XP
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-261 PATCH XPRESS
    50 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 190,00 €
    190,00 €
    1 * 960 ST
       190,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MLBP902-261XP
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-033
    175 µl
    -
    +
    Transparent
    50 racks, 96 each (Econopak)
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 1 570,00 €
    1 570,00 €
    1 * 4.800 ST
       1 570,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MLBP903-033
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
Tecan® Genesis / MiniPrep (black)
Product Information
  • Tip style
    Volume
    Sterile
    Filter
    Tip colour
    Packed
    Availability
    Price
  • 904-251
    1000 µl
    +
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 224,00 €
    224,00 €
    1 * 960 ST
       224,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0061
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 901-252
    20 µl
    -
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 216,00 €
    216,00 €
    1 * 960 ST
       216,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0043
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-253
    50 µl
    -
    -
    Black
    50 racks, 96 each (Econopak)
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 848,00 €
    848,00 €
    1 * 4.800 ST
       848,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0049
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-011G (wide bore)
    175 µl
    +
    +
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 283,00 €
    283,00 €
    1 * 960 ST
       283,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0052
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-251G (wide bore)
    200 µl
    +
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 224,00 €
    224,00 €
    1 * 960 ST
       224,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0054
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-252G (wide bore)
    200 µl
    -
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 206,00 €
    206,00 €
    1 * 960 ST
       206,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0056
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-011G (wide bore)
    1000 µl
    +
    +
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 283,00 €
    283,00 €
    1 * 960 ST
       283,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-251G (wide bore)
    1000 µl
    +
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 224,00 €
    224,00 €
    1 * 960 ST
       224,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0062
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-252G (wide bore)
    1000 µl
    -
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 207,00 €
    207,00 €
    1 * 960 ST
       207,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0064
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-252
    200 µl
    -
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 206,00 €
    206,00 €
    1 * 960 ST
       206,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0055
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-251
    200 µl
    +
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 224,00 €
    224,00 €
    1 * 960 ST
       224,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0053
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-011
    175 µl
    +
    +
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 283,00 €
    283,00 €
    1 * 960 ST
       283,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0051
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-252
    1000 µl
    -
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 206,00 €
    206,00 €
    1 * 960 ST
       206,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0063
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-011
    1000 µl
    +
    +
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 283,00 €
    283,00 €
    1 * 960 ST
       283,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0059
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 901-251
    20 µl
    +
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 239,00 €
    239,00 €
    1 * 960 ST
       239,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0042
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 901-011
    10 µl
    +
    +
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 292,00 €
    292,00 €
    1 * 960 ST
       292,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0041
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-252
    50 µl
    -
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 216,00 €
    216,00 €
    1 * 960 ST
       216,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0048
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-251
    50 µl
    +
    -
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 239,00 €
    239,00 €
    1 * 960 ST
       239,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0047
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 902-011
    50 µl
    +
    +
    Black
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 292,00 €
    292,00 €
    1 * 960 ST
       292,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0046
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 901-253
    20 µl
    -
    -
    Black
    50 racks, 96 each (Econopak)
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 848,00 €
    848,00 €
    1 * 4.800 ST
       848,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0044
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 903-253
    200 µl
    -
    -
    Black
    50 racks, 96 each (Econopak)
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 848,00 €
    848,00 €
    1 * 4.800 ST
       848,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0057
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 904-253
    1000 µl
    -
    -
    Black
    50 racks, 96 each (Econopak)
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 848,00 €
    848,00 €
    1 * 4.800 ST
       848,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0065
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
Tecan® TeMo / Aquarius (transparent)
Product Information
  • Tip style
    Volume
    Sterile
    Filter
    Tip colour
    Packed
    Availability
    Price
  • 908-261
    200 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 131,00 €
    131,00 €
    1 * 960 ST
       131,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1527
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 907-261
    100 µl
    +
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 131,00 €
    131,00 €
    1 * 960 ST
       131,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1528
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 908-021
    150 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 201,00 €
    201,00 €
    1 * 960 ST
       201,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1529
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 908-262
    200 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 128,00 €
    128,00 €
    1 * 960 ST
       128,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1530
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 907-262
    100 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 128,00 €
    128,00 €
    1 * 960 ST
       128,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1531
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 906-262
    50 µl
    -
    -
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 128,00 €
    128,00 €
    1 * 960 ST
       128,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1532
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 906-261
    50 µl
    +
    -
    Transparent
    50 racks, 96 each (Econopak)
    Log in for availability
    Log in for availability
    1 * 4.800 ST
    1 * 4.800 ST - 653,00 €
    653,00 €
    1 * 4.800 ST
       653,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1533
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 906-021
    50 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 201,00 €
    201,00 €
    1 * 960 ST
       201,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1534
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 907-021
    80 µl
    +
    +
    Transparent
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 201,00 €
    201,00 €
    1 * 960 ST
       201,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1535
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact