Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tip refill system, Axygen®

Tip refill system, Axygen®

Supplier:   Corning
Tip refill system, Axygen®
Supplier:  Corning
Certificates
Tip refill system, Axygen®
Tip refill system, Axygen®

2 Options Available Below

Product Details & Documents

The Axygen® Tip Refill System contains durable refill racks that can be easily refilled by hand and are autoclavable.

  • RNase-/DNase-free
  • Non pyrogenic
  • Translucent rack allows tips to be easily seen
  • Colour-coded insert allows for easy identification of tip
Product Family Options

Product Information

  • For use with
    Availability
    Price
  • 732-0958
    Empty refill racks, RFL-R
    Empty refill racks, RFL-R
    10 µl and 200 µl refill inserts
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 39,90 €
    39,90 €
    1 * 10 ST
       39,90 €

  • 732-1403
    Empty refill racks, RFL-R-1200
    Empty refill racks, RFL-R-1200
    1200 µl refill inserts
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52,20 €
    52,20 €
    1 * 10 ST
       52,20 €