Pipette tips, Finntip™ Flex
About this item
New generation of tips offering the user greater flexibility.
- High quality PP, autoclavable
- Ergonomic thin wall tip; low attachment and ejection forces
- Non slip rack with hinged cover
- Easy to use refill system
: Available loose, in racks, in a starter kit (1 rack of 96 and 16 or 20 refills of 96) or a refill kit (16 or 20 refills of 96).
10 ml tips: only available in bags.
22 Options Available Below
Product Details & Documents
New generation of tips offering the user greater flexibility.
- High quality PP, autoclavable
- Ergonomic thin wall tip; low attachment and ejection forces
- Non slip rack with hinged cover
- Easy to use refill system
: Available loose, in racks, in a starter kit (1 rack of 96 and 16 or 20 refills of 96) or a refill kit (16 or 20 refills of 96).
10 ml tips: only available in bags.
Specifications for Product Family
Specifications
- Catalog No:
- 613-0806
- 613-0807
- 613-0809
- 613-0810
- 613-0797
- 613-0798
- 613-0800
- 613-0801
- 613-0190
- 613-5019
- 613-5021
- 613-5022
- 613-0811
- 613-0812
- 613-0814
- 613-0815
- 613-0614
- 613-0615
- 613-0818
- 613-0817
- 613-0819
- 613-1598
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Specifications
Product Family Options
Finntip™ Flex 10 µl
Product Information
- VolumeSterileLengthColour codePackedAvailabilityPrice
- 0,2 - 10 µl-33 mmPink10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 80,00 €80,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST80,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0806Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 0,2 - 10 µl+33 mmPink10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 94,00 €94,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST94,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0807Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 0,2 - 10 µl-33 mmPink20 refills, 96 eachLog in for availabilityLog in for availability1 * 1.920 ST1 * 1.920 ST - 129,00 €129,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.920 ST129,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0809Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 0,2 - 10 µl-33 mmPinkBulkLog in for availabilityLog in for availability1 * 1.000 ST1 * 1.000 ST - 56,00 €56,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.000 ST56,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0810Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ Flex 200 µl
Product Information
- VolumeSterileLengthColour codePackedAvailabilityPrice
- 1 - 200 µl-60 mmYellow10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 73,00 €73,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST73,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0797Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 1 - 200 µl+60 mmYellow10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 90,00 €90,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST90,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0798Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 1 - 200 µl-60 mmYellow20 refills, 96 eachLog in for availabilityLog in for availability1 * 1.920 ST1 * 1.920 ST - 129,00 €129,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.920 ST129,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0800Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 1 - 200 µl-60 mmYellowBulkLog in for availabilityLog in for availability1 * 1.000 ST1 * 1.000 ST - 57,00 €57,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.000 ST57,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0801Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ Flex 300 µl
Product Information
- VolumeSterileLengthColour codePackedAvailabilityPrice
- 5 - 300 µl-61 mmOrange10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 73,00 €73,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST73,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0190Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 5 - 300 µl+61 mmOrange10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 90,00 €90,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST90,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5019Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 5 - 300 µl-61 mmOrange20 refills, 96 eachLog in for availabilityLog in for availability1 * 1.920 ST1 * 1.920 ST - 129,00 €129,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.920 ST129,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5021Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 5 - 300 µl-61 mmOrangeBulkLog in for availabilityLog in for availability1 * 1.000 ST1 * 1.000 ST - 57,00 €57,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.000 ST57,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-5022Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ Flex 1000 µl
Product Information
- VolumeSterileLengthColour codePackedAvailabilityPrice
- 100 - 1000 µl-87 mmBlue10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 76,00 €76,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST76,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0811Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 100 - 1000 µl+87 mmBlue10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 94,00 €94,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST94,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0812Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 100 - 1000 µl-87 mmBlue16 refills, 96 eachLog in for availabilityLog in for availability1 * 1.536 ST1 * 1.536 ST - 118,00 €118,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.536 ST118,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0814Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 100 - 1000 µl-87 mmBlueBulkLog in for availabilityLog in for availability1 * 1.000 ST1 * 1.000 ST - 63,00 €63,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.000 ST63,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0815Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ Flex 1200 µl
Product Information
- VolumeSterileLengthColour codePackedAvailabilityPrice
- 50 - 1200 µl-87 mmTurquoise10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 152,00 €152,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST152,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0614Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 50 - 1200 µl+87 mmTurquoise10 racks, 96 eachLog in for availabilityLog in for availability1 * 960 ST1 * 960 ST - 180,00 €180,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 ST180,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0615Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 50 - 1200 µl-87 mmTurquoiseBulkLog in for availabilityLog in for availability1 * 1.000 ST1 * 1.000 ST - 76,00 €76,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.000 ST76,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0818Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 50 - 1200 µl-87 mmTurquoise16 refills, 96 eachLog in for availabilityLog in for availability1 * 1.536 ST1 * 1.536 ST - 211,00 €211,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 1.536 ST211,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0817Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ Flex 10 ml Extended
Product Information
- VolumeSterileLengthColour codePackedAvailabilityPrice
- 1000 - 10000 µl-267 mm—BulkLog in for availabilityLog in for availability1 * 100 ST1 * 100 ST - 52,00 €52,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 100 ST52,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-0819Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 1000 - 10000 µl+267 mm—BulkLog in for availabilityLog in for availability1 * 50 ST1 * 50 ST - 38,00 €38,00 €
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 50 ST38,00 €Specifications:Low Manufacturing Impact More
Specifications
Cat. No.613-1598Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact