Pipette tips, gel loading, HydroLogix™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Designed for loading vertical electrophoresis gels.
- Choice of round or flat tip
- Available in volumes 1 to 10 µl or 2 to 200 µl
- Intended for loading gels of three thicknesses to suit specific applications
Limited Time Sale
Select items below are currently on sale
Specifications
- Catalog No:
- 732-2256
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)