Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Supplier:   KARTELL
Dropping pipettes
Supplier:  KARTELL
Certificates
Dropping pipettes
Dropping pipettes
2 Options Available Below
Product Details & Documents
PE, graduated.
  • Moulded in one piece
  • High capacity reservoir for take-up and expulsion of liquids
  • Ideal replacement for traditional glass Pasteur pipette and rubber teat
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Length
    Sterile
    Availability
    Price
  • 612-1778
    Dropping pipettes, 1,5 ml, Ø×L 8×134 mm
    Dropping pipettes, 1,5 ml, Ø×L 8×134 mm
    1,5 ml
    8
    134 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 35,90 €
    35,90 €
    1 * 100 ST
       35,90 €
  • 612-1780
    Dropping pipettes, 5,0 ml, Ø×L 10×195 mm
    Dropping pipettes, 5,0 ml, Ø×L 10×195 mm
    5,0 ml
    10 mm
    195 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 53,10 €
    53,10 €
    1 * 100 ST
       53,10 €