Orders LinkContactUsLinkComponent
Transfer pipettes, paddle
Transfer pipettes, paddle
0,2 ml, 50 µl, 124, -, Bag
Catalog # 612-1731
Supplier:   Alpha Laboratories
Transfer pipettes, paddle
Catalog # 612-1731
Supplier:  Alpha Laboratories
Transfer pipettes, paddle
Transfer pipettes, paddle
0,2 ml, 50 µl, 124, -, Bag
Price
59,70 €
Log in for availability
1 * 500 ST
Product Details & Documents
Non toxic, food grade LDPE.
  • For dispensing and mixing applications and popular with diagnostic kit manufacturers
  • Ideal for latex agglutination, pregnancy tests and blood grouping cards
Specifications
  • Packed:
    Bag
  • Sterile:
    -
  • Capacity:
    0,2 ml
  • Length:
    124
  • Drop vol.:
    50 µl