Orders LinkContactUsLinkComponent
Transfer pipettes, fine tip
Transfer pipettes, fine tip
Transfer pipettes, fine tip
Supplier:  Alpha Laboratories
Certificates
Transfer pipettes, fine tip
Transfer pipettes, fine tip
14 Options Available Below
Product Details & Documents
Non toxic, food grade LDPE.
  • For careful control and small volume transfers
  • Ideal for supernatant removal, slide preparation and gel loading
Product Family Options
Product Information
  • Capacity
    Drop vol.
    Length
    Sterile
    Packed
    Availability
    Price
  • ALPCLW4697
    Small bulb fine tip
    Small bulb fine tip
    0,5 ml
    25 µl
    36 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 98,20 €
    98,20 €
    1 * 500 ST
       98,20 €
  • 612-1738
    Ultra micro fine tip
    Ultra micro fine tip
    1,0 ml
    22 µl
    51 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 102,00 €
    102,00 €
    1 * 500 ST
       102,00 €
  • 612-0005
    Extended fine tip, mini
    Extended fine tip, mini
    1,0 ml
    20 µl
    104 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 102,00 €
    102,00 €
    1 * 400 ST
       102,00 €
  • 612-1787
    Extended fine tip, mini
    Extended fine tip, mini
    1,0 ml
    20 µl
    104 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 205,00 €
    205,00 €
    1 * 400 ST
       205,00 €
  • 612-1751
    Extended fine tip
    Extended fine tip
    3,3 ml
    20 µl
    153
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 107,00 €
    107,00 €
    1 * 250 ST
       107,00 €
  • 612-1797
    Extended fine tip
    Extended fine tip
    3,3 ml
    20 µl
    153 mm
    +
    Individually
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 238,00 €
    238,00 €
    1 * 400 ST
       238,00 €
  • ALPCLW4638
    Extended fine tip
    Extended fine tip
    3,3 ml
    20 µl
    153 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 170,00 €
    170,00 €
    1 * 500 ST
       170,00 €
  • 612-0006
    Extended fine tip, standard
    Extended fine tip, standard
    3,3 ml
    20 µl
    157 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 185,00 €
    185,00 €
    1 * 500 ST
       185,00 €
  • 612-1759
    Fine tip
    Fine tip
    3,3 ml
    18 µl
    147 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 118,00 €
    118,00 €
    1 * 500 ST
       118,00 €
  • Fine tip
    3,3 ml
    18 µl
    147 mm
    +
    Individually
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 201,00 €
    201,00 €
    1 * 400 ST
       201,00 €
  • ALPCLW4070
    Fine tip
    Fine tip
    3,3 ml
    18 µl
    147 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 175,00 €
    175,00 €
    1 * 500 ST
       175,00 €
  • Fine tip, maxi
    4,8 ml
    22 µl
    147 mm
    +
    Individually
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 222,00 €
    222,00 €
    1 * 400 ST
       222,00 €
  • ALPCLW4236
    Fine tip, maxi
    Fine tip, maxi
    4,8 ml
    22 µl
    147 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 3.200 ST
    1 * 3.200 ST - 171,00 €
    171,00 €
    1 * 3.200 ST
       171,00 €
  • ALPCLW4636
    Fine tip, maxi
    Fine tip, maxi
    4,8 ml
    22 µl
    147 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 219,00 €
    219,00 €
    1 * 400 ST
       219,00 €