ContactUsLinkComponent
VWR®, Straw Pipettes
  612-1675
undefined
VWR®, Straw Pipettes
  612-1675
  • Packed:
    25 per bag
  • Capacity:
    1 ml
  • Description:
    Straw pipettes
  • Seq:
    01
  • Pk:
    1.000
Straw pipettes made from PP, irradiated, disposable, sterile, without graduation.
  • Traceability including the expiry date on the secondary packaging
  • Zip bag can be pressed closed ensuring product protection
  • Tamper-evident seal guarantees the sterility of the product
 : *The product(s) below are outside the scope of the EU Directive 2019/904