Orders LinkContactUsLinkComponent
Demeter pipettes
Demeter pipettes
Supplier:   HIRSCHMANN
Demeter pipettes
Supplier:  HIRSCHMANN
Certificates
Demeter pipettes
Demeter pipettes
2 Options Available Below
Product Details & Documents
AR-Glas®.
  • Calibrated to deliver 'Ex'
  • Blue graduation
  • Dilution pipettes with two marks at one and one,1 ml or with four marks at 1,0/2,0/2,1/2,2 ml
Length: 250 mm ±5 mm
Length: 250 mm ±5 mm
Certifications: DIN 12838 and DIN 12839
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 1/1,1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • 1,0/2,0/2,1/2,2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,30 €
    13,30 €
    1 * 1 ST
       13,30 €