VWR®, Sterilisation Boxes for Pipettes
Leverancier: VWR Collection
Certificaten
Over dit artikel
Stainless steel, with round base, autoclavable.
- Can be autoclaved at 121 °C and sterilised at 180 °C
- Bottom and underside of the lid are coated in silicone to minimise the risk of damaging the pipettes during transport and sterilisation
Frequently Bought Together