ContactUsLinkComponent
Clamp holders for pipettes, Clamp-ette®
Clamp holders for pipettes, Clamp-ette®
Catalog # 613-3900
Supplier:  HEATHROW
undefined
Clamp holders for pipettes, Clamp-ette®
Catalog # 613-3900
Supplier:  HEATHROW
Specifications
  • For:
    3 pipettes
  • Colour:
    Blue
  • Description:
    Pipettor clamp
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
PP, coloured.
  • Stable, sturdy construction
  • For one to three pipettes of the most widely used brands
  • Secures to tabletops, benches and shelves with a twist of the fingertips
  • Autoclavable