Pressure Bulbs
Supplier: DEUTSCH NEUMANN
Jump to:
About this item
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
5 Options Available Below
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Product Family Options
Atomiser bulb
Product Information
- CapacityAvailabilityPrice
- 612-9952Atomiser bulb with pressure valve, red-brown, size 3, PVCSpecifications:
More
Specifications
Cat. No.612-9952 - DENE4250003Atomiser bulb with suction valve, red-brown, size 3, PVCSpecifications:
More
Specifications
Cat. No.DENE4250003 - DENE4200005Atomiser bulb with pressure valve, red-brown, size 5, PVCAtomiser bulb with pressure valve, red-brown, size 5, PVC102Log in for availability1 * 1 ST24,70 €Specifications:
More
Specifications
Cat. No.DENE4200005
Pressure Ball Set with Net
Product Information
- CapacityAvailabilityPrice
- 612-9953Ball set with net, black, size 5, natural rubber/PVCSpecifications:
More
Specifications
Cat. No.612-9953 - DENE4120003Ball set with net, merlot-red, size 5, natural rubber/PVCBall set with net, merlot-red, size 5, natural rubber/PVC102Log in for availability1 * 1 ST96,70 €Specifications:New Product
More
Specifications
Cat. No.DENE4120003New Product