Orders LinkContactUsLinkComponent
Pipette controller, macro
 :   Brand
Pipette controller, macro
 :  Brand
Pipette controller, macro
Pipette controller, macro
Macro pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
  • Conical silicone adapter offers a secure fit for different diameters
  • Valve system allows easy compression of the bellows
  • Spring-loaded lever enables sensitive filling and delivery of liquids
  • Meniscus is easily adjustable
  • Fully autoclavable at 121 °C (2 bar)
Each pipette controller is supplied with a 3 μm spare membrane filter.
Product Family Options
  • Description
    Colour code
  • Macro pipette controller
    Grey
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €
  • Macro pipette controller
    Green
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €
  • Macro pipette controller
    Blue
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €
  • Macro pipette controller
    Magenta
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €
Accessories
  •  
    Description
    Colour code
  • 612-2202
    Adapter (silicone) Length: 44 mm
    Black
    1 * 1 ST - 5,10 €
  •  
    Description
    Colour code
  • 612-1904
    Valve system (PP, PTFE, silicone)
    Assorted
    1 * 1 ST - 32,00 €
  •  
    Description
    Colour code
  • 612-1965
    Membrane filter 3 µm (PP, PTFE), non sterile
    Assorted
    1 * 1 ST - 7,50 €
  •  
    Description
    Colour code
  • 612-1966
    Membrane filter 3 µm (PP, PTFE), non sterile
    Assorted
    1 * 10 ST - 117,00 €
  •  
    Description
    Colour code
  • 612-1903
    Suction bellows (silicone) with screw ring (PP)
    1 * 1 ST - 22,20 €
  •  
    Description
    Colour code
  • 612-2210
    Adapter support (PP) grey, Length: 49 mm
    Grey
    1 * 1 ST - 4,90 €