Orders LinkContactUsLinkComponent
Pipette controller, micro-classic
Pipette controller, micro-classic
Micro-classic with 1 spare suction tube
Catalog # 612-1912
Supplier:   Brand
Pipette controller, micro-classic
Catalog # 612-1912
Supplier:  Brand
Pipette controller, micro-classic
Pipette controller, micro-classic
Micro-classic with 1 spare suction tube
Price
61,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Suitable for disposable micropipettes with ring mark and other small volume pipettes up to 1 ml (e.g. blood diluting pipettes) with aspiration end diameter maximum 5 mm.
  • Ergonomic shape and simple handling offering comfort and convenience for this job
  • For IVF and medical laboratories
  • Minimises the risk of contamination when working with infectious material
  • Adapter for right- and left-handed operation and suction tube are autoclavable
Specifications
  • Description:
    Micro-classic with 1 spare suction tube
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare adapter with suction tube
    Spare adapter with suction tube
    612-1913
    Log in for availability
    1 * 3 ST - 14,20 €