About this item
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
- Non toxic and inert to biological fluids and most acids
- Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
- Available sterile or non sterile
- Sterility assurance level is SAL 10⁻⁶
Packaging: Sterile pipettes are packed in medical grade paper and non toxic film.
24 Options Available Below
Product Details & Documents
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
- Non toxic and inert to biological fluids and most acids
- Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
- Available sterile or non sterile
- Sterility assurance level is SAL 10⁻⁶
Packaging: Sterile pipettes are packed in medical grade paper and non toxic film.
Documents
Centrifugation Solutions from Avantor VWR® for Liquid Handling
Product Literature
CoC VWR Disposable High Performance Transfer Pipettes.pdf CoC VWR Transfer Pipettes, High-Performance.pdf
Conformity declaration
Product Family Options
General purpose
Product Information
Graduated
Product Information
- CapacityDrops per mlBulb drawStem ØLengthGraduationSterilePackedAvailabilityPrice
Extra large
Product Information
- CapacityDrops per mlBulb drawStem ØLengthGraduationSterilePackedAvailabilityPrice
Narrow stem
Product Information
- CapacityDrops per mlBulb drawStem ØLengthGraduationSterilePackedAvailabilityPrice
Blood bank
Product Information
- CapacityDrops per mlBulb drawStem ØLengthGraduationSterilePackedAvailabilityPrice
Paddle
Product Information
- CapacityDrops per mlBulb drawStem ØLengthGraduationSterilePackedAvailabilityPrice