Orders LinkContactUsLinkComponent
Graduated pipettes, Class AS, Schellbach
Graduated pipettes, Class AS, Schellbach
Supplier:   HIRSCHMANN
Graduated pipettes, Class AS, Schellbach
Supplier:  HIRSCHMANN
Certificates
Graduated pipettes, Class AS, Schellbach
Graduated pipettes, Class AS, Schellbach
8 Options Available Below
Product Details & Documents
AR-Glas®.
  • Calibrated to deliver 'Ex', conformity certified with dated batch identification
  • Marks and inscriptions in blue stain, graduated to the tip, with Schellbach-stripe
  • From 5 ml with a cotton plug upper end
DIN 12697
Length: 360 mm ±5 mm except for 25 ml pipettes: 450 mm ±5 mm.
Certifications: DIN EN ISO 835. Conformity certified (DE-M marking).
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Suppl. to DIN
    Availability
    Price
  • 0,5 ml
    0,01 ml
    0,005 ± ml
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • 1 ml
    0,01 ml
    0,007 ± ml
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 2 ml
    0,02 ml
    0,01 ± ml
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 5 ml
    0,05 ml
    0,03 ± ml
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 5 ml
    0,1 ml
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 10 ml
    0,1 ml
    0,05 ± ml
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 20 ml
    0,1 ml
    0,1 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,90 €
    22,90 €
    1 * 1 ST
       22,90 €
  • 25 ml
    0,1 ml
    0,1 ± ml
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,10 €
    23,10 €
    1 * 1 ST
       23,10 €